Assay Details
| Gene Symbol: RNASE9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110357.1 | NP_001103827.1 | 1 - 2 | 52 | 100 | |
| GenBank mRNA | EF061296.1 | - | 1 - 2 | 52 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the EF061296.1, NM_001110357.1 transcripts.
Set Membership: |
|

RNASE9
Genomic Map