Assay Details
| Gene Symbol: AUTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127231.2 | NP_001120703.1 | 3 - 4 | 1358 | 87 | |
| NM_001127232.2 | NP_001120704.1 | 3 - 4 | 1358 | 87 | ||
| NM_015570.3 | NP_056385.1 | 3 - 4 | 1358 | 87 | ||
| XM_011516010.2 | XP_011514312.1 | 3 - 4 | 1802 | 87 | ||
| XM_011516011.2 | XP_011514313.1 | 3 - 4 | 1802 | 87 | ||
| XM_011516012.2 | XP_011514314.1 | 3 - 4 | 1802 | 87 | ||
| XM_011516013.2 | XP_011514315.1 | 3 - 4 | 1802 | 87 | ||
| XM_011516014.2 | XP_011514316.1 | 3 - 4 | 1802 | 87 | ||
| XM_011516017.2 | XP_011514319.1 | 2 - 3 | 10356 | 87 | ||
| XM_011516018.2 | XP_011514320.1 | 3 - 4 | 337 | 87 | ||
| XM_017011951.1 | XP_016867440.1 | 3 - 4 | 1802 | 87 | ||
| GenBank mRNA | AB007902.2 | - | 5 - 6 | 919 | 87 | |
| AF326917.1 | - | 3 - 4 | 944 | 87 | ||
| BC011643.2 | - | 3 - 4 | 666 | 87 | ||
| BC064693.1 | - | 3 - 4 | 1366 | 30345831 | 87 | |
| JQ670866.1 | - | 3 - 4 | 623 | 87 | ||
More Information
Set Membership: |
|

AUTS2
Genomic Map