Assay Details
| Gene Symbol: AUTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127232.2 | NP_001120704.1 | 4 - 5 | 1391 | 74 | |
| GenBank mRNA | BC011643.2 | - | 4 - 5 | 699 | 74 | |
More Information
Set Membership: |
|

AUTS2
Genomic Map