Assay Details
| Gene Symbol: PAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127366.2 | NP_001120838.1 | 2 - 3 | 708 | 77 | |
| GenBank mRNA | BC101299.3 | - | 2 - 3 | 607 | 40024655 | 77 |
| BC101300.2 | - | 2 - 3 | 607 | 40024656 | 77 | |
| BC101302.2 | - | 2 - 3 | 607 | 40024661 | 77 | |
| BC114363.2 | - | 2 - 3 | 607 | 40024665 | 77 | |
More Information
Set Membership: |
|

PAX3
Genomic Map