Assay Details
| Gene Symbol: BSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128326.2 | NP_001121798.1 | 3 - 4 | 213 | 153 | |
| XM_017026118.1 | XP_016881607.1 | 3 - 4 | 213 | 153 | ||
| GenBank mRNA | DQ227497.1 | - | 3 - 4 | 214 | 153 | |
More Information
Set Membership: |
|

BSPH1
Genomic Map