Assay Details
Gene Symbol: C7orf73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130929.1 | NP_001124401.1 | 1 - 2 | 126 | 104 | |
GenBank mRNA | BC016741.1 | - | 1 - 2 | 90 | 104 | |
BC021734.1 | - | 1 - 2 | 100 | 104 | ||
BC047474.1 | - | 1 - 2 | 95 | 104 | ||
BC107765.1 | - | 1 - 2 | 101 | 104 | ||
DA785733.1 | - | 1 - 2 | 126 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001130929.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
