Assay Details
| Gene Symbol: CACFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135775.2 | NP_001129247.1 | 2 - 3 | 299 | 83 | |
| NM_001242370.1 | NP_001229299.1 | 2 - 3 | 299 | 83 | ||
| XM_011518184.1 | XP_011516486.1 | 2 - 3 | 1041 | 83 | ||
| GenBank mRNA | AK075416.1 | - | 2 - 3 | 297 | 83 | |
| AK298820.1 | - | 2 - 3 | 231 | 83 | ||
| DA699497.1 | - | 2 - 3 | 236 | 83 | ||
More Information
Set Membership: |
|

CACFD1
Genomic Map