Assay Details
| Gene Symbol: SLC22A24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136506.2 | NP_001129978.2 | 9 - 10 | 2037 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A24| Gene Symbol: SLC22A24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136506.2 | NP_001129978.2 | 9 - 10 | 2037 | 88 | |
Set Membership: |
|