Assay Details
| Gene Symbol: ANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142445.1 | NP_001135917.1 | 3 - 4 | 712 | 110 | |
| NM_020476.2 | NP_065209.2 | 41 - 42 | 5708 | 110 | ||
| NM_020477.2 | NP_065210.2 | 41 - 42 | 5222 | 110 | ||
| XM_011544490.2 | XP_011542792.1 | 44 - 45 | 5973 | 110 | ||
| XM_011544494.2 | XP_011542796.1 | 43 - 44 | 5927 | 110 | ||
| XM_011544500.2 | XP_011542802.1 | 41 - 42 | 5810 | 110 | ||
| XM_011544504.2 | XP_011542806.1 | 41 - 42 | 5324 | 110 | ||
| XM_017013319.1 | XP_016868808.1 | 43 - 44 | 5948 | 110 | ||
| XM_017013321.1 | XP_016868810.1 | 44 - 45 | 5917 | 110 | ||
| XM_017013322.1 | XP_016868811.1 | 43 - 44 | 5879 | 110 | ||
| XM_017013323.1 | XP_016868812.1 | 44 - 45 | 5893 | 110 | ||
| XM_017013324.1 | XP_016868813.1 | 42 - 43 | 6006 | 110 | ||
| XM_017013325.1 | XP_016868814.1 | 43 - 44 | 5818 | 110 | ||
| XM_017013327.1 | XP_016868816.1 | 44 - 45 | 5486 | 110 | ||
| XM_017013328.1 | XP_016868817.1 | 43 - 44 | 5441 | 110 | ||
| GenBank mRNA | X16609.1 | - | 41 - 42 | 5708 | 110 | |
More Information
Set Membership: |
|

ANK1
Genomic Map