Assay Details
| Gene Symbol: ZNF780A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142579.1 | NP_001136051.1 | 3 - 4 | 258 | 143 | |
| XM_017026611.1 | XP_016882100.1 | 2 - 3 | 887 | 143 | ||
| XM_017026612.1 | XP_016882101.1 | 3 - 4 | 1026 | 143 | ||
| XM_017026613.1 | XP_016882102.1 | 3 - 4 | 389 | 143 | ||
| XM_017026614.1 | XP_016882103.1 | 3 - 4 | 377 | 143 | ||
| XM_017026615.1 | XP_016882104.1 | 3 - 4 | 331 | 143 | ||
| XM_017026616.1 | XP_016882105.1 | 3 - 4 | 258 | 143 | ||
| GenBank mRNA | AK131328.1 | - | 3 - 4 | 258 | 143 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131328.1, NM_001142579.1, XM_017026611.1, XM_017026612.1, XM_017026613.1, XM_017026614.1, XM_017026615.1, XM_017026616.1 transcripts.
Set Membership: |
|

ZNF780A
Genomic Map