Assay Details
| Gene Symbol: ZNF780A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142579.1 | NP_001136051.1 | 8 - 9 | 749 | 85 | |
| GenBank mRNA | AK131328.1 | - | 8 - 9 | 749 | 85 | |
More Information
Set Membership: |
|

ZNF780A
Genomic Map