Assay Details
| Gene Symbol: LIPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144869.1 | NP_001138341.1 | 1 - 2 | 470 | 71 | |
| GenBank mRNA | CX752266.1 | - | 1 - 2 | 452 | 71 | |
More Information
Set Membership: |
|

LIPT2
Genomic Map