Assay Details
| Gene Symbol: GLYCTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144951.1 | NP_001138423.1 | 3 - 4 | 625 | 75 | |
| GenBank mRNA | AY295075.1 | - | 2 - 3 | 558 | 75 | |
More Information
Set Membership: |
|

GLYCTK
Genomic Map