Assay Details
| Gene Symbol: BRI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159491.1 | NP_001152963.1 | 1 - 2 | 270 | 103 | |
| NM_015379.4 | NP_056194.1 | 1 - 2 | 270 | 103 | ||
| XM_017011936.1 | XP_016867425.1 | 1 - 2 | 264 | 103 | ||
| GenBank mRNA | AB055977.1 | - | 1 - 2 | 262 | 103 | |
| AF041430.2 | - | 1 - 2 | 239 | 103 | ||
| AF106966.1 | - | 1 - 2 | 221 | 103 | ||
| BC018737.1 | - | 1 - 2 | 262 | 4875257 | 103 | |
| BC062370.1 | - | 1 - 2 | 234 | 6598528 | 103 | |
| BF108440.1 | - | 1 - 2 | 116 | 103 | ||
More Information
Set Membership: |
|

BRI3
Genomic Map