Assay Details
| Gene Symbol: BRI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159491.1 | NP_001152963.1 | 2 - 3 | 378 | 90 | |
| GenBank mRNA | BF108440.1 | - | 2 - 3 | 224 | 90 | |
| BF432881.1 | - | 2 - 3 | 223 | 90 | ||
More Information
Set Membership: |
|

BRI3
Genomic Map