Assay Details
| Gene Symbol: ACOT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160094.1 | NP_001153566.1 | 2 - 3 | 602 | 105 | |
| GenBank mRNA | AK309738.1 | - | 2 - 3 | 467 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001160094.1 transcripts.
Set Membership: |
|

ACOT13
Genomic Map