Assay Details
| Gene Symbol: PDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161779.1 | NP_001155251.1 | 2 - 3 | 310 | 67 | |
| GenBank mRNA | DA769567.1 | - | 2 - 3 | 310 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001161779.1 transcripts.
Set Membership: |
|

PDP1
Genomic Map