Assay Details
| Gene Symbol: ANKRD66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162435.2 | NP_001155907.2 | 1 - 2 | 77 | 73 | |
| XM_017010148.1 | XP_016865637.1 | 1 - 2 | 65 | 73 | ||
| GenBank mRNA | DC417416.1 | - | 1 - 2 | 77 | 73 | |
More Information
Set Membership: |
|

ANKRD66
Genomic Map