Assay Details
| Gene Symbol: ANKRD66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162435.2 | NP_001155907.2 | 4 - 5 | 558 | 71 | |
| GenBank mRNA | AK304342.1 | - | 4 - 5 | 513 | 71 | |
| BU608132.1 | - | 1 - 2 | 516 | 71 | ||
| DC417416.1 | - | 4 - 5 | 558 | 71 | ||
More Information
Set Membership: |
|

ANKRD66
Genomic Map