Assay Details
| Gene Symbol: ZNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164527.1 | NP_001157999.1 | 1 - 2 | 67 | 80 | |
| NM_001164529.1 | NP_001158001.1 | 1 - 2 | 67 | 80 | ||
| NM_001164530.1 | NP_001158002.1 | 1 - 2 | 67 | 80 | ||
| NM_007134.1 | NP_009065.1 | 1 - 2 | 67 | 80 | ||
| XM_005259211.1 | XP_005259268.1 | 1 - 2 | 67 | 80 | ||
| GenBank mRNA | AK098011.1 | - | 1 - 2 | 67 | 80 | |
| AL833637.1 | - | 1 - 2 | 25 | 80 | ||
| BX647775.1 | - | 1 - 2 | 35 | 80 | ||
| DA232862.1 | - | 1 - 2 | 60 | 80 | ||
| DB117846.1 | - | 1 - 2 | 67 | 80 | ||
More Information
Set Membership: |
|

ZNF135
Genomic Map