Assay Details
| Gene Symbol: DHX40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166301.1 | NP_001159773.1 | 16 - 17 | 2117 | 117 | |
| NM_024612.4 | NP_078888.4 | 17 - 18 | 2348 | 117 | ||
| XM_006722083.2 | XP_006722146.1 | 16 - 17 | 2213 | 117 | ||
| XM_011525253.2 | XP_011523555.1 | 16 - 17 | 2111 | 117 | ||
| XM_011525254.2 | XP_011523556.1 | 17 - 18 | 2277 | 117 | ||
| XM_017025080.1 | XP_016880569.1 | 16 - 17 | 2112 | 117 | ||
| XM_017025081.1 | XP_016880570.1 | 16 - 17 | 2196 | 117 | ||
| XM_017025082.1 | XP_016880571.1 | 16 - 17 | 2164 | 117 | ||
| XM_017025083.1 | XP_016880572.1 | 16 - 17 | 2161 | 117 | ||
| GenBank mRNA | AF260270.1 | - | 16 - 17 | 2416 | 117 | |
| AF461690.1 | - | 17 - 18 | 2348 | 117 | ||
| AK025713.1 | - | 7 - 8 | 820 | 117 | ||
| AK091334.1 | - | 13 - 14 | 1640 | 117 | ||
| AK095681.1 | - | 17 - 18 | 2261 | 117 | ||
| AK123683.1 | - | 2 - 3 | 2287 | 117 | ||
| AK225268.1 | - | 7 - 8 | 824 | 117 | ||
| AK225344.1 | - | 7 - 8 | 820 | 117 | ||
| AK226048.1 | - | 17 - 18 | 2226 | 117 | ||
| AK299151.1 | - | 16 - 17 | 2030 | 117 | ||
| AL832510.1 | - | 13 - 14 | 1566 | 117 | ||
| BC024187.2 | - | 17 - 18 | 2263 | 4822080 | 117 | |
More Information
Set Membership: |
|

DHX40
Genomic Map