Assay Details
| Gene Symbol: DHX40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166301.1 | NP_001159773.1 | 2 - 3 | 340 | 124 | |
| GenBank mRNA | AK299151.1 | - | 2 - 3 | 253 | 124 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK299151.1 transcripts.
Set Membership: |
|

DHX40
Genomic Map