Assay Details
| Gene Symbol: ALG13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168385.2 | NP_001161857.1 | 4 - 5 | 519 | 109 | |
| NR_033125.2 | 4 - 5 | 415 | 109 | |||
| GenBank mRNA | AL524204.3 | - | 4 - 5 | 338 | 109 | |
| BE747995.1 | - | 4 - 5 | 448 | 109 | ||
More Information
Set Membership: |
|

ALG13
Genomic Map