Assay Details
| Gene Symbol: GOLGA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174124.1 | NP_001167595.1 | 4 - 5 | 444 | 80 | |
| GenBank mRNA | AK026613.1 | - | 4 - 5 | 424 | 80 | |
More Information
Set Membership: |
|

GOLGA7
Genomic Map