Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178100.1 | NP_001171571.1 | 3 - 4 | 555 | 99 | |
| GenBank mRNA | BC100912.2 | - | 3 - 4 | 546 | 40003723 | 99 |
More Information
Set Membership: |
|

CD8B
Genomic Map