Assay Details
| Gene Symbol: PPP1R3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184745.1 | NP_001171674.1 | 2 - 3 | 154 | 77 | |
| XM_005272687.3 | XP_005272744.1 | 2 - 3 | 1115 | 77 | ||
| XM_017029933.1 | XP_016885422.1 | 2 - 3 | 5522 | 77 | ||
| XM_017029935.1 | XP_016885424.1 | 2 - 3 | 1115 | 77 | ||
| GenBank mRNA | AK056297.1 | - | 1 - 2 | 514 | 77 | |
| AK303738.1 | - | 3 - 4 | 420 | 77 | ||
| BC131588.1 | - | 2 - 3 | 127 | 77 | ||
| DC412626.1 | - | 3 - 4 | 422 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK056297.1, NM_001184745.1 transcripts.
Set Membership: |
|

PPP1R3F
Genomic Map