Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190977.2 | NP_001177906.1 | 1 - 2 | 213 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190977.2 transcripts.
Set Membership: |
|

YAF2
Genomic Map