Assay Details
Gene Symbol: C6orf99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195032.1 | NP_001181961.1 | 1 - 2 | 101 | 75 | |
GenBank mRNA | BF688381.1 | - | 1 - 2 | 102 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195032.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
