Assay Details
Gene Symbol: CLRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195794.1 | NP_001182723.1 | 3 - 4 | 764 | 84 | |
NR_046380.2 | 4 - 5 | 915 | 84 | |||
GenBank mRNA | HM626132.1 | - | 3 - 4 | 473 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
