Assay Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198595.1 | NP_001185524.1 | 2 - 3 | 165 | 86 | |
| GenBank mRNA | CR936807.1 | - | 2 - 3 | 148 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198595.1 transcripts.
Set Membership: |
|

STON1
Genomic Map