Assay Details
| Gene Symbol: RBM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198844.1 | NP_001185773.1 | 2 - 3 | 556 | 82 | |
| GenBank mRNA | AK303984.1 | - | 2 - 3 | 481 | 82 | |
More Information
Set Membership: |
|

RBM4
Genomic Map