Assay Details
| Gene Symbol: YY1AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198900.1 | NP_001185829.1 | 2 - 3 | 249 | 102 | |
| GenBank mRNA | BC044887.1 | - | 2 - 3 | 246 | 5276742 | 102 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC044887.1, NM_001198900.1 transcripts.
Set Membership: |
|

YY1AP1
Genomic Map