Assay Details
| Gene Symbol: MINOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204082.1 | NP_001191011.1 | 1 - 2 | 127 | 74 | |
| GenBank mRNA | BM784596.1 | - | 1 - 2 | 80 | 74 | |
More Information
Set Membership: |
|

MINOS1
Genomic Map