Assay Details
| Gene Symbol: USMG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206426.1 | NP_001193355.1 | 1 - 2 | 103 | 59 | |
| GenBank mRNA | AJ272056.1 | - | 1 - 2 | 73 | 59 | |
| AK311888.1 | - | 1 - 2 | 43 | 59 | ||
| AL110185.1 | - | 1 - 2 | 42 | 59 | ||
| BC072683.1 | - | 1 - 2 | 50 | 6056764 | 59 | |
| BC091507.1 | - | 1 - 2 | 48 | 30417704 | 59 | |
| DW446355.1 | - | 1 - 2 | 119 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ272056.1, AK311888.1, BC072683.1, BC091507.1, NM_001206426.1 transcripts.
Set Membership: |
|

USMG5
Genomic Map