Assay Details
| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004329.2 | NP_001004329.2 | 1 - 2 | 576 | 88 | |
| GenBank mRNA | AK131230.1 | - | 1 - 2 | 576 | 88 | |
More Information
Set Membership: |
|

DBX2
Genomic Map