Assay Details
| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004329.2 | NP_001004329.2 | 2 - 3 | 668 | 103 | |
| GenBank mRNA | AK131230.1 | - | 2 - 5 | 668 | 103 | |
More Information
Set Membership: |
|

DBX2
Genomic Map