Assay Details
Gene Symbol: OR5K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004736.3 | NP_001004736.2 | 1 | 1013 | 129 | |
GenBank mRNA | BC136918.1 | - | 1 | 1013 | 9020909 | 129 |
BC136919.1 | - | 1 | 1013 | 9020910 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
