Assay Details
| Gene Symbol: NCMAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010980.4 | NP_001010980.1 | 1 - 2 | 89 | 119 | |
| GenBank mRNA | CB961010.1 | - | 1 - 2 | 117 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001010980.4 transcripts.
Set Membership: |
|

NCMAP
Genomic Map