Assay Details
| Gene Symbol: NCMAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010980.4 | NP_001010980.1 | 3 - 4 | 265 | 71 | |
| XM_005245872.3 | XP_005245929.1 | 3 - 4 | 1603 | 71 | ||
| XM_011541463.2 | XP_011539765.1 | 3 - 4 | 1062 | 71 | ||
| GenBank mRNA | AK124519.1 | - | 3 - 4 | 230 | 71 | |
| BC050474.1 | - | 3 - 4 | 337 | 71 | ||
| BC127785.1 | - | 2 - 3 | 167 | 40134460 | 71 | |
| BC147014.1 | - | 2 - 3 | 186 | 9056942 | 71 | |
| BC147015.1 | - | 2 - 3 | 186 | 9056943 | 71 | |
| CB961010.1 | - | 3 - 4 | 293 | 71 | ||
More Information
Set Membership: |
|

NCMAP
Genomic Map