Assay Details
| Gene Symbol: WDR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242414.1 | NP_001229343.1 | 2 - 3 | 333 | 88 | |
| XM_011537345.1 | XP_011535647.1 | 2 - 3 | 286 | 88 | ||
| GenBank mRNA | AK090978.1 | - | 2 - 3 | 274 | 88 | |
| AL832242.1 | - | 2 - 3 | 272 | 88 | ||
More Information
Set Membership: |
|

WDR20
Genomic Map