Assay Details
| Gene Symbol: RSPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242910.1 | NP_001229839.1 | 5 - 6 | 1147 | 67 | |
| GenBank mRNA | DQ165085.1 | - | 5 - 6 | 889 | 67 | |
More Information
Set Membership: |
|

RSPO1
Genomic Map