Assay Details
| Gene Symbol: PQLC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243729.1 | NP_001230658.1 | 3 - 4 | 402 | 89 | |
| GenBank mRNA | BC144066.1 | - | 3 - 4 | 369 | 89 | |
| BG217991.1 | - | 2 - 3 | 236 | 89 | ||
More Information
Set Membership: |
|

PQLC2L
Genomic Map