Assay Details
| Gene Symbol: APH1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243772.1 | NP_001230701.1 | 2 - 3 | 544 | 63 | |
| NR_045035.1 | 2 - 3 | 680 | 63 | |||
| GenBank mRNA | AK298832.1 | - | 2 - 3 | 351 | 63 | |
| AK298876.1 | - | 2 - 3 | 459 | 63 | ||
More Information
Set Membership: |
|

APH1A
Genomic Map