Assay Details
| Gene Symbol: TPD52L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243895.1 | NP_001230824.1 | 5 - 6 | 621 | 62 | |
| GenBank mRNA | DA547106.1 | - | 5 - 6 | 522 | 62 | |
More Information
Set Membership: |
|

TPD52L2
Genomic Map