Assay Details
| Gene Symbol: FAM19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252216.1 | NP_001239145.1 | 1 - 2 | 216 | 69 | |
| GenBank mRNA | AY325114.1 | - | 1 - 2 | 216 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY325114.1, NM_001252216.1 transcripts.
Set Membership: |
|

FAM19A1
Genomic Map