Assay Details
| Gene Symbol: STPG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004353.3 | NP_001004353.2 | 1 - 2 | 141 | 59 | |
| NM_001256699.1 | NP_001243628.1 | 1 - 2 | 141 | 59 | ||
| NM_001256700.1 | NP_001243629.1 | 1 - 2 | 141 | 59 | ||
| NM_001256701.1 | NP_001243630.1 | 1 - 2 | 141 | 59 | ||
| NR_046339.1 | 1 - 2 | 141 | 59 | |||
| NR_046340.1 | 1 - 2 | 141 | 59 | |||
| XM_006717113.2 | XP_006717176.1 | 2 - 3 | 287 | 59 | ||
| XM_006717115.2 | XP_006717178.1 | 1 - 2 | 144 | 59 | ||
| GenBank mRNA | AK097565.1 | - | 1 - 2 | 124 | 59 | |
| BC033939.1 | - | 1 - 2 | 121 | 5272537 | 59 | |
| BC050094.1 | - | 1 - 2 | 97 | 59 | ||
| BC132726.1 | - | 1 - 2 | 141 | 40146748 | 59 | |
| BC144308.2 | - | 1 - 2 | 141 | 9052828 | 59 | |
| BC144311.1 | - | 1 - 2 | 141 | 9052831 | 59 | |
| BC144314.1 | - | 1 - 2 | 141 | 9052834 | 59 | |
| DB453255.1 | - | 1 - 2 | 108 | 59 | ||
More Information
Set Membership: |
|

STPG3
Genomic Map