Assay Details
| Gene Symbol: STPG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256700.1 | NP_001243629.1 | 4 - 5 | 514 | 72 | |
| NM_001256701.1 | NP_001243630.1 | 4 - 5 | 514 | 72 | ||
| GenBank mRNA | BC033939.1 | - | 4 - 5 | 494 | 5272537 | 72 |
More Information
Set Membership: |
|

STPG3
Genomic Map