Assay Details
| Gene Symbol: STPG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256699.1 | NP_001243628.1 | 5 - 6 | 822 | 92 | |
| NM_001256700.1 | NP_001243629.1 | 5 - 6 | 657 | 92 | ||
| NR_046340.1 | 5 - 6 | 565 | 92 | |||
| XM_006717113.2 | XP_006717176.1 | 6 - 7 | 968 | 92 | ||
| XM_006717115.2 | XP_006717178.1 | 5 - 6 | 648 | 92 | ||
| GenBank mRNA | BC033939.1 | - | 5 - 6 | 637 | 5272537 | 92 |
| BC132726.1 | - | 5 - 6 | 822 | 40146748 | 92 | |
| BC144314.1 | - | 5 - 6 | 822 | 9052834 | 92 | |
More Information
Set Membership: |
|

STPG3
Genomic Map