Assay Details
| Gene Symbol: FAAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256945.1 | NP_001243874.1 | 1 - 2 | 86 | 80 | |
| NM_001256947.1 | NP_001243876.1 | 1 - 2 | 86 | 80 | ||
| NR_046427.1 | 1 - 2 | 86 | 80 | |||
| XM_017000557.1 | XP_016856046.1 | 1 - 2 | 80 | |||
| GenBank mRNA | BU192678.1 | - | 1 - 2 | 47 | 80 | |
| CN397640.1 | - | 1 - 2 | 65 | 80 | ||
More Information
Set Membership: |
|

FAAP20
Genomic Map