Assay Details
| Gene Symbol: COX14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257133.1 | NP_001244062.1 | 2 - 3 | 378 | 111 | |
| GenBank mRNA | DW418935.1 | - | 2 - 3 | 171 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257133.1 transcripts.
Set Membership: |
|

COX14
Genomic Map